Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028399.4 | NP_001023570.2 | 2 - 3 | 478 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |