Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074376.2 | NP_506777.2 | 5 - 6 | 568 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |