Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDNRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031632.1 | NP_001026802.1 | 3 - 4 | 738 | 70 | |
XM_005628830.3 | XP_005628887.1 | 4 - 5 | 935 | 70 | ||
XM_014119229.2 | XP_013974704.1 | 4 - 5 | 934 | 70 | ||
GenBank mRNA | AB183284.2 | - | 3 - 4 | 738 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Canis familiaris Probe spans exons |