Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031819.1 | NP_001026989.1 | 3 - 4 | 1252 | 71 | |
GenBank mRNA | AY916758.1 | - | 3 - 4 | 1252 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Canis familiaris Probe spans exons |