Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020808.1 | NP_001018644.1 | 2 - 3 | 222 | 77 | |
XM_005627072.3 | XP_005627129.1 | 3 - 4 | 489 | 77 | ||
XM_022425734.1 | XP_022281442.1 | 3 - 4 | 501 | 77 | ||
XM_022425735.1 | XP_022281443.1 | 3 - 4 | 353 | 77 | ||
XM_022425736.1 | XP_022281444.1 | 3 - 4 | 266 | 77 | ||
XM_022425738.1 | XP_022281446.1 | 2 - 3 | 177 | 77 | ||
GenBank mRNA | DQ002813.1 | - | 2 - 3 | 222 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Inventoried Probe spans exons |