Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195142.1 | NP_001182071.1 | 6 - 7 | 383 | 77 | |
NM_001195143.1 | NP_001182072.1 | 6 - 7 | 383 | 77 | ||
XM_005615754.3 | XP_005615811.2 | 6 - 7 | 436 | 77 | ||
XM_005615755.3 | XP_005615812.2 | 6 - 7 | 492 | 77 | ||
XM_014114177.2 | XP_013969652.2 | 6 - 7 | 677 | 77 | ||
XM_014114183.2 | XP_013969658.1 | 6 - 7 | 587 | 77 | ||
XM_014114188.2 | XP_013969663.1 | 6 - 7 | 538 | 77 | ||
GenBank mRNA | M63934.1 | - | 6 - 7 | 360 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |