Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003220.1 | NP_001003220.1 | 4 - 5 | 581 | 75 | |
GenBank mRNA | Z81018.1 | - | 581 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |