Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_860510.5 | XP_865603.1 | 3 - 4 | 612 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |