Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618876.2 | XP_005618933.1 | 1 - 2 | 104 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |