Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAICS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639458.4 | XP_003639506.1 | 8 - 9 | 1113 | 84 | |
XM_022427005.1 | XP_022282713.1 | 21 - 22 | 6014 | 84 | ||
XM_022427006.1 | XP_022282714.1 | 20 - 21 | 5906 | 84 | ||
XM_022427008.1 | XP_022282716.1 | 24 - 25 | 6407 | 84 | ||
XM_022427009.1 | XP_022282717.1 | 21 - 22 | 5916 | 84 | ||
XM_022427010.1 | XP_022282718.1 | 20 - 21 | 5699 | 84 | ||
XM_022427011.1 | XP_022282719.1 | 23 - 24 | 6271 | 84 | ||
XM_022427012.1 | XP_022282720.1 | 19 - 20 | 5633 | 84 | ||
XM_022427013.1 | XP_022282721.1 | 22 - 23 | 6106 | 84 | ||
XM_022427015.1 | XP_022282723.1 | 14 - 15 | 2421 | 84 | ||
XM_022427016.1 | XP_022282724.1 | 14 - 15 | 2323 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |