Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003434684.4 | XP_003434732.1 | 435 | 85 | ||
XM_005620570.3 | XP_005620627.1 | 459 | 85 | |||
XM_005620571.3 | XP_005620628.1 | 425 | 85 | |||
XM_014113950.2 | XP_013969425.1 | 458 | 85 | |||
XM_844929.5 | XP_850022.1 | 492 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris |