Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625143.3 | XP_005625200.1 | 3 - 4 | 372 | 75 | |
XM_005625144.3 | XP_005625201.1 | 3 - 4 | 354 | 75 | ||
XM_022423633.1 | XP_022279341.1 | 3 - 4 | 247 | 75 | ||
XM_537799.6 | XP_537799.3 | 3 - 4 | 420 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |