Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417626.1 | XP_022273334.1 | 12 - 13 | 1159 | 80 | |
XM_022417627.1 | XP_022273335.1 | 12 - 13 | 1148 | 80 | ||
XM_536412.6 | XP_536412.2 | 11 - 12 | 1056 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |