Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615589.3 | XP_005615646.1 | 1 - 2 | 298 | 63 | |
XM_014113304.2 | XP_013968779.1 | 1 - 2 | 298 | 63 | ||
XM_541214.5 | XP_541214.3 | 1 - 2 | 297 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |