Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639250.3 | XP_005639307.1 | 4 - 5 | 558 | 117 | |
XM_005639251.3 | XP_005639308.1 | 4 - 5 | 567 | 117 | ||
XM_545016.5 | XP_545016.2 | 4 - 5 | 570 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Probe spans exons |