Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637325.2 | XP_005637382.1 | 1 - 2 | 145 | 62 | |
XM_014108492.2 | XP_013963967.1 | 1 - 2 | 145 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |