Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617283.3 | XP_005617340.1 | 691 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris |