Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALML6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620406.3 | XP_005620463.1 | 610 | 90 | ||
XM_005620410.3 | XP_005620467.1 | 471 | 90 | |||
XM_022418914.1 | XP_022274622.1 | 558 | 90 | |||
XM_022418915.1 | XP_022274623.1 | 546 | 90 | |||
XM_022418916.1 | XP_022274624.1 | 603 | 90 | |||
XM_022418917.1 | XP_022274625.1 | 599 | 90 | |||
XM_022418918.1 | XP_022274626.1 | 571 | 90 | |||
XM_022418919.1 | XP_022274627.1 | 523 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris |