Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048130.1 | NP_001041595.1 | 3 - 4 | 331 | 83 | |
XM_005638180.3 | XP_005638237.1 | 4 - 5 | 487 | 83 | ||
XM_005638181.2 | XP_005638238.1 | 4 - 5 | 511 | 83 | ||
XM_022412499.1 | XP_022268207.1 | 3 - 4 | 284 | 83 | ||
GenBank mRNA | DQ494380.1 | - | 3 - 4 | 331 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |