Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NREP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003638950.4 | XP_003638998.1 | 274 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris |