Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR89A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630713.3 | XP_005630770.1 | 2 - 3 | 268 | 83 | |
XM_022404930.1 | XP_022260638.1 | 2 - 3 | 270 | 83 | ||
XM_022404931.1 | XP_022260639.1 | 2 - 3 | 268 | 83 | ||
XM_844483.5 | XP_849576.1 | 2 - 3 | 268 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Canis familiaris Probe spans exons |