Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011888.1 | NP_001011888.1 | 3 | 296 | 139 | |
GenBank mRNA | AJ875418.1 | - | 3 | 296 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Amplicon spans exons and probe does not span exons |