Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626916.3 | XP_005626973.1 | 3 | 499 | 114 | |
XM_005626917.3 | XP_005626974.1 | 3 | 556 | 114 | ||
XM_022424932.1 | XP_022280640.1 | 2 | 164 | 114 | ||
XM_022424933.1 | XP_022280641.1 | 3 | 391 | 114 | ||
XM_532031.6 | XP_532031.3 | 3 | 445 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |