Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022406692.1 | XP_022262400.1 | 3 | 961 | 61 | |
XM_022406693.1 | XP_022262401.1 | 3 | 957 | 61 | ||
XM_022406694.1 | XP_022262402.1 | 3 | 900 | 61 | ||
XM_022406695.1 | XP_022262403.1 | 3 | 957 | 61 | ||
XM_022406696.1 | XP_022262404.1 | 2 | 811 | 61 | ||
XM_022406697.1 | XP_022262405.1 | 2 | 807 | 61 | ||
XM_022406698.1 | XP_022262406.1 | 3 | 896 | 61 | ||
XM_022406699.1 | XP_022262407.1 | 2 | 750 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |