Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635683.3 | XP_005635740.1 | 4 | 1385 | 68 | |
XM_005635685.3 | XP_005635742.1 | 3 | 1349 | 68 | ||
XM_005635686.3 | XP_005635743.1 | 4 | 1483 | 68 | ||
XM_014107600.2 | XP_013963075.1 | 3 | 2336 | 68 | ||
XM_014107601.2 | XP_013963076.1 | 3 | 2434 | 68 | ||
XM_014107602.2 | XP_013963077.1 | 3 | 1447 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |