Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621949.2 | XP_005622006.1 | 4 | 2446 | 114 | |
XM_022420523.1 | XP_022276231.1 | 3 | 2531 | 114 | ||
XM_022420524.1 | XP_022276232.1 | 4 | 2455 | 114 | ||
XM_022420525.1 | XP_022276233.1 | 3 | 2288 | 114 | ||
XM_022420526.1 | XP_022276234.1 | 2 | 1898 | 114 | ||
XM_547288.5 | XP_547288.2 | 3 | 2350 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |