Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_132651.1 | 845 | 87 | |||
GenBank mRNA | DN864176.1 | - | 593 | 87 | ||
EF191197.1 | - | 845 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |