Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244115.1 | NP_001231044.1 | 2 - 3 | 517 | 70 | |
GenBank mRNA | AY272060.1 | - | 2 - 3 | 517 | 70 | |
JI874714.1 | - | 2 - 3 | 508 | 70 | ||
JP056454.1 | - | 2 - 3 | 508 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |