Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244849.1 | NP_001231778.1 | 6 - 7 | 1016 | 86 | |
GenBank mRNA | AF046870.1 | - | 6 - 7 | 1016 | 86 | |
JI889270.1 | - | 6 - 7 | 781 | 86 | ||
JP061104.1 | - | 6 - 7 | 781 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |