Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lix1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007631673.1 | XP_007629863.1 | 4 - 5 | 641 | 95 | |
GenBank mRNA | JI866069.1 | - | 2 - 3 | 162 | 95 |