Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502141.2 | XP_003502189.1 | 2 - 3 | 272 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |