Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509596.1 | XP_003509644.1 | 4 - 5 | 552 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |