Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503627.2 | XP_003503675.1 | 3 - 4 | 1848 | 61 | |
XM_007644213.1 | XP_007642403.1 | 4 - 5 | 588 | 61 | ||
XM_007644214.1 | XP_007642404.1 | 4 - 5 | 447 | 61 | ||
XM_007644215.1 | XP_007642405.1 | 3 - 4 | 352 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |