Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509272.2 | XP_003509320.1 | 8 - 9 | 1198 | 108 | |
GenBank mRNA | JI891023.1 | - | 2 - 3 | 164 | 109 | |
JP038792.1 | - | 2 - 3 | 164 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |