Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il4i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651465.1 | XP_007649655.1 | 3 - 4 | 446 | 89 | |
XM_007651466.1 | XP_007649656.1 | 3 - 4 | 446 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Detect Genomic DNA Probe spans exons |