Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505814.2 | XP_003505862.1 | 3 - 4 | 829 | 67 | |
GenBank mRNA | JI870251.1 | - | 3 - 4 | 659 | 67 | |
JP057033.1 | - | 3 - 4 | 659 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |