Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643211.1 | XP_007641401.1 | 5 - 6 | 637 | 64 | |
XM_007643212.1 | XP_007641402.1 | 5 - 6 | 1364 | 64 | ||
XM_007643213.1 | XP_007641403.1 | 6 - 7 | 874 | 64 | ||
GenBank mRNA | JI883595.1 | - | 5 - 6 | 678 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |