Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007633623.1 | XP_007631813.1 | 5 - 6 | 686 | 105 | |
GenBank mRNA | JI874837.1 | - | 1 - 2 | 114 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |