Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496800.2 | XP_003496848.1 | 2 - 3 | 272 | 104 | |
GenBank mRNA | JI876866.1 | - | 2 - 3 | 272 | 104 | |
JP044741.1 | - | 2 - 3 | 272 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Probe spans exons |