Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642567.1 | XP_007640757.1 | 4 - 5 | 667 | 79 | |
GenBank mRNA | JI882104.1 | - | 2 - 3 | 122 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |