Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fsip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503397.1 | XP_003503445.1 | 23 - 24 | 20558 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |