Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680257.3 | XP_005680314.1 | 4 - 5 | 877 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |