Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285570.1 | NP_001272499.1 | 3 - 4 | 328 | 92 | |
XM_013965265.2 | XP_013820719.1 | 3 - 4 | 351 | 92 | ||
GenBank mRNA | DQ010419.1 | - | 3 - 4 | 328 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |