Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696604.3 | XP_005696661.1 | 1 - 2 | 1066 | 99 |
Set Membership: |
![]() ![]() ![]() ![]() |