Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO579962.1 | - | 7 - 8 | 1412 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |