Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675458.3 | XP_005675515.2 | 5 - 6 | 779 | 97 | |
XM_005675459.3 | XP_005675516.2 | 5 - 6 | 868 | 97 | ||
XM_018049095.1 | XP_017904584.1 | 4 - 5 | 685 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |