Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLIRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686168.3 | XP_005686225.2 | 1 - 2 | 125 | 74 | |
GenBank mRNA | JR119186.1 | - | 236 | |||
JR119187.1 | - | 236 |
Set Membership: |
Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |