Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMERB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697522.2 | XP_005697579.2 | 5 - 6 | 671 | 88 | |
GenBank mRNA | JO634954.2 | - | 1 - 2 | 1305 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |