Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685487.2 | XP_005685544.1 | 3 - 4 | 535 | 88 | |
XM_005685488.3 | XP_005685545.1 | 3 - 4 | 674 | 88 | ||
GenBank mRNA | JI861612.1 | - | 2 - 3 | 403 | 88 | |
JO403893.2 | - | 1 - 2 | 957 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |