Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRICKLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038734.1 | XP_017894223.1 | 6 - 7 | 2369 | 92 | |
XM_018038735.1 | XP_017894224.1 | 7 - 8 | 2477 | 92 |
Gene Symbol: TOMM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038736.1 | XP_017894225.1 | 2 - 3 | 296 | 92 | |
GenBank mRNA | JO367710.2 | - | 1 - 2 | 312 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |